Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pentanone p-Toluenesulfonylhydrazone 98.0+%, TCI America™
SDP

Catalog No. P18135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18135G Supplier TCI America Supplier No. P18135G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28495-72-9
Molecular Formula C12H18N2O2S
Molecular Weight (g/mol) 254.348
InChI Key GUEMDXLIHCONOI-UHFFFAOYSA-N
Synonym 3-Pentanone Tosylhydrazone
PubChem CID 277909
IUPAC Name 4-methyl-N-(pentan-3-ylideneamino)benzenesulfonamide
SMILES CCC(=NNS(=O)(=O)C1=CC=C(C=C1)C)CC

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 5 g
Formula Weight 254.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Pentanone p-Toluenesulfonylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.