accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH2C

CAS RN 2851-95-8

IUPAC Name: 1-ethenyl-2-methyl-1H-imidazole
Synonyms: 1-ethenyl-2-methylimidazole
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: BDHGFCVQWMDIQX-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results