Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-vinylimidazole 97.0+%, TCI America™
SDP

Catalog No. M07531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M07531G Supplier TCI America Supplier No. M07531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2851-95-8
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00037893
InChI Key BDHGFCVQWMDIQX-UHFFFAOYSA-N
PubChem CID 76109
IUPAC Name 1-ethenyl-2-methylimidazole
SMILES CC1=NC=CN1C=C

Specifications

Color Yellow
Boiling Point 94°C
Quantity 1 g
UN Number 3267
Formula Weight 108.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-vinylimidazole
EINECSNumber (5)-0384
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.