accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONNN

CAS RN 28539-02-8

IUPAC Name: (1H-1,2,3-benzotriazol-1-yl)methanol
Synonyms: 1,2,3-benzotriazol-1-ylmethanol
Molecular Weight (g/mol): 149.15
Molecular Formula: C7H7N3O
InChi Key: MXJIHEXYGRXHGP-UHFFFAOYSA-N
SMILES: OCN1N=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results