Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Benzotriazole-1-methanol 98.0+%, TCI America™
SDP

Catalog No. B190325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B190325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B190325G Supplier TCI America Supplier No. B190325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28539-02-8
Molecular Formula C7H7N3O
Molecular Weight (g/mol) 149.153
MDL Number MFCD00179118
InChI Key MXJIHEXYGRXHGP-UHFFFAOYSA-N
Synonym 1-(Hydroxymethyl)-1H-benzotriazole
PubChem CID 224169
IUPAC Name benzotriazol-1-ylmethanol
SMILES C1=CC=C2C(=C1)N=NN2CO

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 149.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1H-Benzotriazole-1-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.