accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2855-08-5

IUPAC Name: 1-chloro-3,3-dimethylbutane
Synonyms: 1-chloro-3,3-dimethylbutane
Molecular Weight (g/mol): 120.62
Molecular Formula: C6H13Cl
InChi Key: XGCKOSFYXBAPQM-UHFFFAOYSA-N
SMILES: CC(C)(C)CCCl
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results