Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3,3-dimethylbutane, 97%

Catalog No. AAL1175318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1175318 50 g
AAL1175309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1175318 Supplier Thermo Scientific Chemicals Supplier No. L1175318
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2855-08-5 | C6H13Cl | 120.62 g/mol

1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2855-08-5
MDL Number MFCD00018980

Specifications

Density 0.867
Boiling Point 116°C to 118°C
Flash Point 7°C (44°F)
Molecular Formula C14H12N4O4
Refractive Index 1.417
Linear Formula (CH3)3CCH2CH2Cl
Quantity 50 g
UN Number UN1993
Beilstein 1731465
Synonym m-tolualdehyde 2,4-dinitrophenylhydrazone, m-tolualdehyde, 2,4-dinitrophenyl hydrazone, benzaldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, 3'-methylbenzaldehyde 2,4-dinitrophenylhydrazone, m-tolualdehyd-2,4-dinitrophenylhydrazone, m-tolualdehyde 2,4-dinitro-phenylhydrazone, 3-methylbenzaldehyde 2,4-dinitrophenyl hydrazone #, 1-2,4-dinitrophenyl-2-3-methylbenzylidene hydrazine, n-e-3-methylphenyl methylideneamino-2,4-dinitroaniline, m-tolualdehyde 2,4-dinitrophenylhydrazone, environmental standard
Solubility Information Not miscible or difficult to mix in water.
InChI Key PQBRXQUGVASQBU-UHFFFAOYSA-N
SMILES CC1=CC=CC(=C1)C=NNC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name N-[(E)-(3-methylphenyl)methylideneamino]-2,4-dinitroaniline
Molecular Weight (g/mol) 300.27
PubChem CID 9604108
Formula Weight 120.62
Percent Purity 97%
Chemical Name or Material 1-Chloro-3,3-dimethylbutane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 220-665-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.