accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28562-53-0

CH3OOOHN

CAS RN 28562-53-0

IUPAC Name: 4-oxoazetidin-2-yl acetate
Synonyms: 4-oxoazetidin-2-yl acetate
Molecular Weight (g/mol): 129.12
Molecular Formula: C5H7NO3
InChi Key: OEYMQQDJCUHKQS-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results