Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetoxy-2-azetidinone, 97%, Thermo Scientific Chemicals

Catalog No. AC456640010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456640010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456640010 Supplier Thermo Scientific Chemicals Supplier No. 456640010

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28562-53-0
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00010593
InChI Key OEYMQQDJCUHKQS-UHFFFAOYNA-N
Synonym 4-acetoxy-2-azetidinone, 2-azetidinone, 4-acetyloxy, 4-acetoxyazetidinone, 4-oxoazetidin-2-yl acetate, 2-oxoazetidinium 4-acetate, 2-azetidinone, 4-acetyloxy-, 4r, 4-acetoxy-azetidinone, acmc-20bsq9, 4-acetoxyazetidin-2-one, 4-acetoxyazetidine-2-one
PubChem CID 119981
IUPAC Name (4-oxoazetidin-2-yl) acetate
SMILES CC(=O)OC1CC(=O)N1

Specifications

Melting Point 35.0°C to 38.0°C
Elemental Analysis 44.65 to 48.37% (C) 10.41 to 11.28% (N)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.12
Percent Purity 97%
Chemical Name or Material 4-Acetoxy-2-azetidinone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 249-083-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.