accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2859-67-8

IUPAC Name: 3-(pyridin-3-yl)propan-1-ol
Synonyms: 3-pyridinepropanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ZUGAIMFLQLPTKB-UHFFFAOYSA-N
SMILES: OCCCC1=CC=CN=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)