Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinepropanol 98.0+%, TCI America™
SDP

Catalog No. P035010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P035010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P035010G Supplier TCI America Supplier No. P035010G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2859-67-8
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00006414
InChI Key ZUGAIMFLQLPTKB-UHFFFAOYSA-N
Synonym 3-(3-Hydroxypropyl)pyridine, 3-(3-Pyridyl)propanol
PubChem CID 17861
IUPAC Name 3-pyridin-3-ylpropan-1-ol
SMILES C1=CC(=CN=C1)CCCO

Specifications

Color Yellow
Boiling Point 133°C
Quantity 10 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pyridinepropanol
RTECSNumber UT7390000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.