accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNClOHNOFFFFFF

CAS RN 285986-88-1

IUPAC Name: 2-({[3,5-bis(trifluoromethyl)phenyl]formamido}oxy)-N-(4-chlorophenyl)propanamide
Synonyms: 2-({[3,5-bis(trifluoromethyl)phenyl]formamido}oxy)-N-(4-chlorophenyl)propanamide
Molecular Weight (g/mol): 454.75
Molecular Formula: C18H13ClF6N2O3
InChi Key: DSMXVSGJIDFLKP-UHFFFAOYNA-N
SMILES: CC(ONC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(=O)NC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results