Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCG-1423 98.0+%, TCI America™
SDP

Catalog No. C344925MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C344925MG 25 mg
C3449100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C344925MG Supplier TCI America Supplier No. C344925MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 285986-88-1
Molecular Formula C18H13ClF6N2O3
Molecular Weight (g/mol) 454.753
MDL Number MFCD01566719
InChI Key DSMXVSGJIDFLKP-UHFFFAOYSA-N
Synonym N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)benzamide
PubChem CID 2726015
IUPAC Name N-[1-(4-chloroanilino)-1-oxopropan-2-yl]oxy-3,5-bis(trifluoromethyl)benzamide
SMILES CC(C(=O)NC1=CC=C(C=C1)Cl)ONC(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F

Specifications

Color White-Yellow
Quantity 25 mg
Formula Weight 454.75
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material CCG-1423
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.