accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SBrH3CN

CAS RN 28599-52-2

IUPAC Name: 5-bromo-2,4-dimethyl-1,3-thiazole
Synonyms: 5-bromo-2,4-dimethyl-1,3-thiazole
Molecular Weight (g/mol): 192.07
Molecular Formula: C5H6BrNS
InChi Key: BSFCVAYFJQLZEU-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results