accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFCH3H3CNNCH3H3C

CAS RN 286014-34-4

IUPAC Name: 1,3-bis(propan-2-yl)-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1,3-diisopropylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 240.05
Molecular Formula: C9H17BF4N2
InChi Key: PSKQPXYUPOKHPY-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC(C)N1C=C[N+](=C1)C(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)