Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diisopropylimidazolium Tetrafluoroborate 96.0+%, TCI America™
SDP

Catalog No. D36231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36231G Supplier TCI America Supplier No. D36231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 286014-34-4
Molecular Formula C9H17BF4N2
Molecular Weight (g/mol) 240.053
MDL Number MFCD08276804
InChI Key PSKQPXYUPOKHPY-UHFFFAOYSA-N
Synonym IiPr.HBF4
PubChem CID 16218102
IUPAC Name 1,3-di(propan-2-yl)imidazol-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC(C)N1C=C[N+](=C1)C(C)C

Specifications

Melting Point 62-79°C
Quantity 1 g
UN Number 1759
Formula Weight 240.05
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diisopropylimidazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.