accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NHNN(Z)(Z)

CAS RN 28631-66-5

IUPAC Name: 2-methyl-4-{[4-(phenylamino)phenyl][4-(phenylimino)cyclohexa-2,5-dien-1-ylidene]methyl}aniline
Synonyms: water blue
Molecular Weight (g/mol): 453.59
Molecular Formula: C32H27N3
InChi Key: LBLFVSANTXZPEX-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1N)C(C1=CC=C(NC2=CC=CC=C2)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1
Color 
  • (3)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Grade 
  • (1)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)