Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline Blue (Water Soluble/Certified Biological Stain), Fisher Chemical

Catalog No. A96725 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A967-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A967-25

missing translation for 'validMainframeMsg'

For use in histology.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28631-66-5
Molecular Formula C32H27N3
Molecular Weight (g/mol) 453.59
MDL Number MFCD00036130
InChI Key LBLFVSANTXZPEX-UHFFFAOYSA-N
Synonym Acid Blue 93, Methyl Blue, Acid Blue, Soluble Blue
PubChem CID 131675892
IUPAC Name 2-methyl-4-{[4-(phenylamino)phenyl][4-(phenylimino)cyclohexa-2,5-dien-1-ylidene]methyl}aniline
SMILES CC1=CC(=CC=C1N)C(C1=CC=C(NC2=CC=CC=C2)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1

Specifications

Bio. Stain Comm. Certification Pass Test
Dye Content Report mL of 0.1N TiCL3/g
Formula Weight 799.8
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 42780
Color Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Aniline Blue
Safety handling Check Alert storage code Red
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin.
NFPA
Health:2 Flammability:0 Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.