accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONH2

CAS RN 28657-75-2

IUPAC Name: 1-(6-amino-2H-1,3-benzodioxol-5-yl)ethan-1-one
Synonyms: 1-(6-amino-2H-1,3-benzodioxol-5-yl)ethanone
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: DWTHYSZSRJOMSC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=C(OCO2)C=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results