accessibility menu, dialog, popup

UserName

Viewing profile as user:
ISFFFFF

CAS RN 286947-67-9

IUPAC Name: 1-iodo-3-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-iodo-3-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 330.05
Molecular Formula: C6H4F5IS
InChi Key: OPPXUPGOUVHFHM-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(I)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results