Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodophenylsulfur Pentafluoride 93.0+%, TCI America™
SDP

Catalog No. I06941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06941G Supplier TCI America Supplier No. I06941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 286947-67-9
Molecular Formula C6H4F5IS
Molecular Weight (g/mol) 330.05
MDL Number MFCD03788513
InChI Key OPPXUPGOUVHFHM-UHFFFAOYSA-N
PubChem CID 2779197
IUPAC Name 1-iodo-3-(pentafluoro-λ⁶-sulfanyl)benzene
SMILES FS(F)(F)(F)(F)C1=CC(I)=CC=C1

Specifications

Boiling Point 57°C
Quantity 1 g
Formula Weight 330.05
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Iodophenylsulfur Pentafluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.