CAS RN 287196-57-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ γ-Secretase Inhibitor VI
The γ-Secretase Inhibitor VI, also referenced under CAS 287196-57-0, controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.