accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2873-74-7

IUPAC Name: pentanedioyl dichloride
Synonyms: glutaryl chloride
Molecular Weight (g/mol): 169
Molecular Formula: C5H6Cl2O2
InChi Key: YVOFTMXWTWHRBH-UHFFFAOYSA-N
SMILES: ClC(=O)CCCC(Cl)=O
Boiling Point 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)