Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaryl dichloride, 97%

Catalog No. AC119991000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119991000 100 mL Glass bottle
AC119990250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC119991000 Supplier Thermo Scientific Chemicals Supplier No. 119991000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2873-74-7 | C5H6Cl2O2 | 169.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2873-74-7
Molecular Formula C5H6Cl2O2
Molecular Weight (g/mol) 169.01
MDL Number MFCD00000755
InChI Key YVOFTMXWTWHRBH-UHFFFAOYSA-N
Synonym glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride
PubChem CID 17887
IUPAC Name pentanedioyl dichloride
SMILES C(CC(=O)Cl)CC(=O)Cl

Specifications

Color Brown-Red or Colorless to Yellow
Density 1.3240g/mL
Boiling Point 216.0°C to 218.0°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4720 to 1.4760
Linear Formula ClCO(CH2)3COCl
Quantity 100 mL
Beilstein 02, 634
Specific Gravity 1.324
Solubility Information Solubility in water: reacts with water.
Formula Weight 169.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Glutaryl dichloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
EINECSNumber 220-711-1
RTECSNumber SA2583000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.