accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2875-18-5

IUPAC Name: 2,3,5,6-tetrafluoropyridine
Synonyms: 2,3,5,6-tetrafluoropyridine
Molecular Weight (g/mol): 151.06
Molecular Formula: C5HF4N
InChi Key: HWIPMBCMGVXOKN-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)N=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results