Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluoropyridine 98.0+%, TCI America™
SDP

Catalog No. T17785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17785G 5 g
T17781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17785G Supplier TCI America Supplier No. T17785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2875-18-5
Molecular Formula C5HF4N
Molecular Weight (g/mol) 151.06
MDL Number MFCD00011734
InChI Key HWIPMBCMGVXOKN-UHFFFAOYSA-N
Synonym pyridine, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-pyridine, 2,3,5,6-tetrafluoropyridine, pubchem9303, acmc-209h4j, ksc494q4b, hwipmbcmgvxokn-uhfffaoysa, 2,3,5,6-tetrafluoro-pyridine; pyridine, 2,3,5,6-tetrafluoro
PubChem CID 137749
IUPAC Name 2,3,5,6-tetrafluoropyridine
SMILES FC1=CC(F)=C(F)N=C1F

Specifications

Color Yellow
Boiling Point 101°C
Quantity 5 g
UN Number 1993
Formula Weight 151.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.