accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2876-08-6

H3CNH3CN

CAS RN 2876-08-6

IUPAC Name: 1,2-dimethyl-1H-1,3-benzodiazole
Synonyms: 1,2-dimethyl-1,3-benzodiazole
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: PJQIBTFOXWGAEN-UHFFFAOYSA-N
SMILES: CN1C(C)=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results