Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethylbenzimidazole 98.0+%, TCI America™
SDP

Catalog No. D36655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36655G 5 g
D36651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36655G Supplier TCI America Supplier No. D36655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2876-08-6
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00022840
InChI Key PJQIBTFOXWGAEN-UHFFFAOYSA-N
PubChem CID 312693
IUPAC Name 1,2-dimethylbenzimidazole
SMILES CC1=NC2=CC=CC=C2N1C

Specifications

Melting Point 111°C
Boiling Point 164°C
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethylbenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.