accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2876-78-0

IUPAC Name: methyl 2-(naphthalen-1-yl)acetate
Synonyms: methyl 2-(naphthalen-1-yl)acetate
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: YGGXZTQSGNFKPJ-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results