Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-Naphthaleneacetate 98.0+%, TCI America™
SDP

Catalog No. N057025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N057025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N057025G Supplier TCI America Supplier No. N057025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2876-78-0
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00004045
InChI Key YGGXZTQSGNFKPJ-UHFFFAOYSA-N
Synonym methyl 1-naphthaleneacetate, methyl 1-naphthylacetate, hormonit, kartofin, methyl 2-naphthalen-1-yl acetate, 1-naphthaleneacetic acid methyl ester, m 1 growth regulator, 1-naphthaleneacetic acid, methyl ester, methyl alpha-naphthylacetate, methyl naphthaleneacetic acid
PubChem CID 17891
IUPAC Name methyl 2-naphthalen-1-ylacetate
SMILES COC(=O)CC1=CC=CC2=CC=CC=C21

Specifications

Color Colorless
Boiling Point 195°C
Quantity 25 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 1-Naphthaleneacetate
RTECSNumber QJ1225000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.