CAS RN 287917-96-8
IUPAC Name:
4-bromo-1-methyl-1H-pyrazole-3-carbaldehyde
Synonyms:
4-bromo-1-methylpyrazole-3-carbaldehyde
Molecular Weight (g/mol):
189.01
Molecular Formula:
C5H5BrN2O
InChi Key:
KGIYMMPLYIITLL-UHFFFAOYSA-N
SMILES:
CN1C=C(Br)C(C=O)=N1
1
–
1
of
1
results
1
Promotions Available
4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
CAS: 287917-96-8 Molecular Formula: C5H5BrN2O Molecular Weight (g/mol): 189.01 InChI Key: KGIYMMPLYIITLL-UHFFFAOYSA-N Synonym: 4-bromo-1-methyl-1h-pyrazole-3-carbaldehyde,4-bromo-3-formyl-1-methyl-1h-pyrazole,4-bromo-1-methylpyrazole-3-carboxaldehyde,4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde,1h-pyrazole-3-carboxaldehyde, 4-bromo-1-methyl,pubchem23668,acmc-209h4p,buttpark 88\08-06,4-bromo-3-formyl-1-methylpyrazole,1h-pyrazole-3-carboxaldehyde,4-bromo-1-methyl PubChem CID: 2735595 IUPAC Name: 4-bromo-1-methylpyrazole-3-carbaldehyde SMILES: CN1C=C(C(=N1)C=O)Br