Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AC447430050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447430050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC447430050 Supplier Thermo Scientific Chemicals Supplier No. 447430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 287917-96-8 | Molecular Formula: C5H5BrN2O | Molecular Weight: 189.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287917-96-8
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.01
InChI Key KGIYMMPLYIITLL-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carbaldehyde, 4-bromo-3-formyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazole-3-carboxaldehyde, 4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde, 4-bromo-1-methyl, pubchem23668, acmc-209h4p, buttpark 88\08-06, 4-bromo-3-formyl-1-methylpyrazole, 1h-pyrazole-3-carboxaldehyde,4-bromo-1-methyl
PubChem CID 2735595
IUPAC Name 4-bromo-1-methylpyrazole-3-carbaldehyde
SMILES CN1C=C(C(=N1)C=O)Br

Specifications

Melting Point 71°C to 76°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 189.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.