CAS RN 2880-58-2
IUPAC Name:
2-methoxycyclohexa-2,5-diene-1,4-dione
Synonyms:
2-methoxy-p-benzoquinone
Molecular Weight (g/mol):
138.12
Molecular Formula:
C7H6O3
InChi Key:
ZJKWJHONFFKJHG-UHFFFAOYSA-N
SMILES:
COC1=CC(=O)C=CC1=O
1
–
1
of
1
results
1
Methoxybenzoquinone 99.0+%, TCI America™
CAS: 2880-58-2 Molecular Formula: C7H6O3 Molecular Weight (g/mol): 138.122 MDL Number: MFCD00144755 InChI Key: ZJKWJHONFFKJHG-UHFFFAOYSA-N PubChem CID: 76146 IUPAC Name: 2-methoxycyclohexa-2,5-diene-1,4-dione SMILES: COC1=CC(=O)C=CC1=O