Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxybenzoquinone 99.0+%, TCI America™
SDP

Catalog No. M10455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10455G 5 g
M10451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10455G Supplier TCI America Supplier No. M10455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2880-58-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00144755
InChI Key ZJKWJHONFFKJHG-UHFFFAOYSA-N
PubChem CID 76146
IUPAC Name 2-methoxycyclohexa-2,5-diene-1,4-dione
SMILES COC1=CC(=O)C=CC1=O

Specifications

Melting Point 147°C
Color Yellow
Quantity 5 g
Formula Weight 138.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methoxybenzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.