accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOOCH3

CAS RN 28821-18-3

IUPAC Name: 7-(ethylamino)-4-methyl-2H-chromen-2-one
Synonyms: 7-(ethylamino)-4-methylchromen-2-one
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: OTNIKUTWXUODJZ-UHFFFAOYSA-N
SMILES: CCNC1=CC=C2C(C)=CC(=O)OC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results