Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-(Ethylamino)-4-methylcoumarin 98.0+%, TCI America™
SDP

Catalog No. E1129200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1129200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1129200MG Supplier TCI America Supplier No. E1129200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28821-18-3
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00467040
InChI Key OTNIKUTWXUODJZ-UHFFFAOYSA-N
Synonym Coumarin 445
PubChem CID 120061
IUPAC Name 7-(ethylamino)-4-methylchromen-2-one
SMILES CCNC1=CC2=C(C=C1)C(=CC(=O)O2)C

Specifications

Melting Point 154°C
Color Yellow
Quantity 200 mg
Formula Weight 203.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-(Ethylamino)-4-methylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.