accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 288246-16-2

IUPAC Name: 4-bromo-1H-pyrazole-5-carbonitrile
Synonyms: 4-bromo-2H-pyrazole-3-carbonitrile
Molecular Weight (g/mol): 171.99
Molecular Formula: C4H2BrN3
InChi Key: ZXVFKQRZKKGVNJ-UHFFFAOYSA-N
SMILES: BrC1=C(NN=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results