CAS RN 288259-39-2
IUPAC Name:
N-(5-aminopentyl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide; trifluoroacetic acid
Synonyms:
N-(5-aminopentyl)-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide; trifluoroacetic acid
Molecular Weight (g/mol):
442.5
Molecular Formula:
C17H29F3N4O4S
InChi Key:
QWKKJNXOPDNIEU-UHFFFAOYNA-N
SMILES:
OC(=O)C(F)(F)F.NCCCCCNC(=O)CCCCC1SCC2NC(=O)NC12
1
–
1
of
1
results
1
N-(5-Aminopentyl)biotinamide trifluoroacetate salt
CAS: 288259-39-2 Molecular Formula: C17H29F3N4O4S Molecular Weight (g/mol): 442.50 MDL Number: MFCD03452814 InChI Key: QWKKJNXOPDNIEU-UHFFFAOYNA-N Synonym: n-5-aminopentyl biotinamide trifluoroacetate salt,biotin-cadaverine,n-5-aminopentyl biotinamide trifluoroacetate salt, solid PubChem CID: 16007074