Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(5-Aminopentyl)biotinamide trifluoroacetate salt

Catalog No. AAJ66382MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66382MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66382MF Supplier Thermo Scientific Chemicals Supplier No. J66382MF
Add to Cart

CAS: 288259-39-2 | C17H29F3N4O4S | 442.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288259-39-2
Molecular Formula C17H29F3N4O4S
Molecular Weight (g/mol) 442.50
MDL Number MFCD03452814
InChI Key QWKKJNXOPDNIEU-UHFFFAOYNA-N
Synonym n-5-aminopentyl biotinamide trifluoroacetate salt, biotin-cadaverine, n-5-aminopentyl biotinamide trifluoroacetate salt, solid
PubChem CID 16007074

Specifications

Color White
Quantity 50 mg
Sensitivity Light and moisture sensitive
Formula Weight 442.5
Physical Form Solid
Chemical Name or Material N-(5-Aminopentyl)biotinamide trifluoroacetate salt
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.