accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2883-45-6

IUPAC Name: hepta-1,6-dien-4-ol
Synonyms: hepta-1,6-dien-4-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: UTGFOWQYZKTZTN-UHFFFAOYSA-N
SMILES: OC(CC=C)CC=C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)