Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,6-Heptadien-4-ol

Catalog No. 1115895G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-158-95G 5 g
11-158-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-158-95G Supplier Sigma Aldrich Supplier No. 1115895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,6-heptadien-4-ol can help in synthesizing the derivatives of guanine, adenine, uracil and thymine via mitsunobu condensation.1,6-heptadien-4-ol (monoterpene alcohol) was used as the internal standard in the quantitation and identification of the important aroma components in wine
  • It was used to study the co- and terpolymerization reactions of carbon monoxide and propene with dicationic biphosphine palladium(ii) as the catalyst

Specifications

CAS 2883-45-6
Density 0.864 g/mL (at 25°C (literature))
Boiling Point 151°C (lit.)
Molecular Formula C7H12O
Refractive Index n20/D 1.45 (literature)
Linear Formula H2C=CHCH2CH(OH)CH2CH=CH2
MDL Number MFCD00008663
Quantity 5 g
Molecular Weight (g/mol) 112.17
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.