accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOF(E/Z)

CAS RN 288401-62-7

IUPAC Name: 6-(2,5-dihydro-1,2-oxazol-5-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
Synonyms: 4-fluoro-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H6FNO2
InChi Key: AOHLNWMWBVBSIK-UHFFFAOYSA-N
SMILES: FC1=CC(=C2ONC=C2)C(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results