Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-(5-isoxazolyl)phenol 98.0+%, TCI America™
SDP

Catalog No. F10495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10495G 5 g
F10491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10495G Supplier TCI America Supplier No. F10495G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 288401-62-7
Molecular Formula C9H6FNO2
Molecular Weight (g/mol) 179.15
MDL Number MFCD03070563
InChI Key AOHLNWMWBVBSIK-UHFFFAOYSA-N
Synonym 5-(5-Fluoro-2-hydroxyphenyl)isoxazole
PubChem CID 6816866
IUPAC Name 4-fluoro-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNO2)C=C1F

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 5 g
Formula Weight 179.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-(5-isoxazolyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.