accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2885-00-9

IUPAC Name: octadecane-1-thiol
Synonyms: 1-octadecanethiol
Molecular Weight (g/mol): 286.56
Molecular Formula: C18H38S
InChi Key: QJAOYSPHSNGHNC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCS
Percent Purity 
  • (2)
  • (2)
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)