Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Octadecanethiol

Catalog No. 503126196 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6196 100 mL
50-312-5918 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-6196 Supplier Sigma Aldrich Supplier No. O1858100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Octadecanethiol

Specifications

CAS 2885-00-9
Melting Point 30°C to 33°C
Density 0.847 g/mL (at 25°C)
Boiling Point 204°C to 210°C (11 mmHg)
Molecular Formula C18H38S
Linear Formula CH3(CH2)17 SH
MDL Number MFCD00004886
Quantity 100 mL
Synonym Mercaptan C18; Octadecyl mercaptan; Stearyl mercaptan
Molecular Weight (g/mol) 286.56
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.