accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOONHH3CCH2

CAS RN 288617-75-4

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methyldec-9-enoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methyldec-9-enoic acid
Molecular Weight (g/mol): 421.54
Molecular Formula: C26H31NO4
InChi Key: MADFVGMQNXRFAF-SANMLTNESA-N
SMILES: CC(CCCCCCC=C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results