Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Grubbs Catalyst™ M207

Catalog No. 6823652G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-236-52G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 68-236-52G Supplier Sigma Aldrich Supplier No. 6823652G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A general purpose olefin metathesis catalyst similar in reactivity to Grubbs Catalyst™ 2nd Generation.Grubbs Catalyst™ Technology for Olefin Metathesis by AldrichProduct of UmicoreProduct LicenseThis product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer's use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.For any further information on product please refer to your local Umicore PMC contact

Specifications

CAS 288617-75-4
Molecular Formula C26H31NO4
Linear Formula C26H31NO4
Quantity 5 g
Synonym (S)-N-Fmoc-alpha-(7-Octenyl)alanine; Fmoc-(S)-2-(7-octenyl)alanine; Fmoc-(S)-2-amino-2-methyl-dec-6-enoic acid
Molecular Weight (g/mol) 421.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.