accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOONHH3CCH2

CAS RN 288617-77-6

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylhept-6-enoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylhept-6-enoic acid
Molecular Weight (g/mol): 379.46
Molecular Formula: C23H25NO4
InChi Key: MRJFPZWLOJOINV-HSZRJFAPSA-N
SMILES: CC(CCCC=C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results