accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COO(E)

CAS RN 28897-58-7

IUPAC Name: (2E)-2-methylhex-2-enoate
Synonyms: (2E)-2-methylhex-2-enoate
Molecular Weight (g/mol): 127.16
Molecular Formula: C7H11O2
InChi Key: AKOVMBAFZSPEQU-AATRIKPKSA-M
SMILES: CCC\C=C(/C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results