Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-hexenoic Acid, TCI America™
SDP

Catalog No. M114710ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M114710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M114710ML Supplier TCI America Supplier No. M114710ML

Chemical Identifiers

CAS 28897-58-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015855
InChI Key AKOVMBAFZSPEQU-AATRIKPKSA-N
Synonym 2-methyl-2-hexenoic acid, 2-hexenoic acid, 2-methyl, e-2-methylhex-2-enoic acid, 2-methylhex-2-enoic acid, e-2-methyl-2-hexenoic acid, 2e-2-methylhex-2-enoic acid, 2-methyl-2e-hexenoic acid, 2e-2-methyl-2-hexenoic acid #
PubChem CID 5282650
IUPAC Name (E)-2-methylhex-2-enoic acid
SMILES CCCC=C(C)C(=O)O

Specifications

Color Yellow
Boiling Point 89°C
Quantity 10 mL
UN Number 3265
Formula Weight 128.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-hexenoic Acid
EINECSNumber (2)-0609
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.